missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius Arium™ Advance L Pre-filter
Arium™ Advance L Pre-filter protects and increases the running life of the downstream reverse osmosis module.
Marca: Sartorius H2O-CPFL-1
Descrizione
- The Arium™ Advance L 5 μm pre-filter protects and increases the running life of the downstream reverse osmosis module.
- The filter reliably removes suspended particles and particle contamination from the feed water.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifica
Filter made of polypropylene with 5 μm separation rate | |
Reverse Osmosis (Type 3) | |
11 x 18 x 26 cm |
Potable Tap Water | |
Pre-filter |