Learn More
ATG2A Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 23226-1-AP-150UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Required for both autophagosome formation and regulation of lipid droplet morphology and dispersion.Specifica
| ATG2A | |
| Polyclonal | |
| Unconjugated | |
| ATG2A | |
| ATG2A, KIAA0404 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 23130 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.35 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q2TAZ0 | |
| ATG2A | |
| ATG2A Fusion Protein Ag19709 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |