missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATP5C1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
140.00€ - 399.00€
Specifica
Antigene | ATP5C1 |
---|---|
Concentrazione | 0.32 mg/mL |
Applicazioni | Western Blot |
Classificazione | Polyclonal |
Coniugato | Unconjugated |
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
16807613
|
Proteintech
10910-1-AP-20UL |
20 μL |
140.00€
20µL |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
16897603
|
Proteintech
10910-1-AP-150UL |
150 μL |
399.00€
150µL |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
Descrizione
This gene encodes a subunit of mitochondrial ATP synthase. Mitochondrial ATP synthase catalyzes ATP synthesis, utilizing an electrochemical gradient of protons across the inner membrane during oxidative phosphorylation. ATP synthase is composed of two linked multi-subunit complexes: the soluble catalytic core, F1, and the membrane-spanning component, Fo, comprising the proton channel. The catalytic portion of mitochondrial ATP synthase consists of 5 different subunits (alpha, beta, gamma, delta, and epsilon) assembled with a stoichiometry of 3 alpha, 3 beta, and a single representative of the other 3. The proton channel consists of three main subunits (a, b, c). This gene encodes the gamma subunit of the catalytic core. Alternatively spliced transcript variants encoding different isoforms have been identified. This gene also has a pseudogene on chromosome 14.Specifica
ATP5C1 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Rat, Mouse | |
P35435, P36542, Q91VR2 | |
116550, 11949, 509 | |
ATP5C1 Fusion Protein Ag1343 | |
Primary | |
-20°C | |
ATP5C1 |
0.32 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
ATP5C, ATP5C1, ATP5CL1, ATPG, F ATPase gamma subunit | |
ATP5C1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |