Learn More
3M™ Aura™ Particulate Respirator
Particulate respirator with 3-panel design allows for greater facial movement
Brand:  3M™ 7000088725
 View more versions of this product
Product Code. 17247620
Questo articolo non è attualmente disponibile o è stato dismesso.
La preghiamo di consultare la pagina del prodotto per individuare le possibili alternative.
View alternative products »
Explore more special offers
This item is not returnable. View return policy
Description
Description
  • Original 3-panel design allows for greater facial movement
  • Low-resistance filter technology helps you breathe more easily
  • Sculpted upper panel conforms to the nose and contours of the face for an effective seal
  • Embossed top panel helps reduce eyewear fogging
  • Chin tab designed for ease of positioning and adjustment
  • Easy to store away with flat-fold design and individual wrapping
  • Colour-coded headbands for easy identification: yellow for FFP1 (NFP 4)
  • Sculpted nose panel helps conform to the nose and contours of the face and helps to improve compatibility with 3M eyewear
  • Maximum usage level: Up to 4 x TLV
  • 3M™ Cool Flow™ Valve helps reduce heat build-up in hot and humid conditions
  • Available in large or small pack sizes
TRUSTED_SUSTAINABILITY
Specifications
Specifications
FFP1
9310+
Particulate Respirator
No
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.