Learn More
Thermo Scientific™ Automation Tips for PerkinElmer™/Packard™ Robotic Liquid Handling Systems with 96- and 384-Channel MDT Dispense Heads
Sfoglia Tutto MBP Molecular BioProducts Prodotti
Descrizione
Increase pipetting effectiveness with these automation tips, achieving accurate and precise performance due to superior tip straightness and a low coefficient of variation.
Thermo Scientific™ Automation Tips for Perkin Elmer™/Packard™ Robotic Liquid Handling Systems with Varispan™ Dispense Arms, which undergo a rigorous 15-point quality control program to ensure consistency. This leads to accurate and precise performance due to superior tip straightness and a low coefficient of variation. The pipette tips are validated on the actual workstations for which they are created and the sterile pipette tips are certified free of RNase, DNase, DNA, endotoxin, bioburden and pyrogen.
- Available in non-sterile, sterile, with Thermo Scientific™ ART™ filter technology and in low retention
- Increased precision for the toughest multi-dispensing assays
- Low percentage coefficient of variation (%CV)
Specifica
Specifica
| Volume (sistema metrico) | 30 μL |
| Filtrato | Non-filtered |
| Sterilità | Sterile |
| Colore | Trasparente |
| Autoclavabile | Not Autoclavable |
| Certificazione/Conformità | ISO 9001 and ISO 13485 |
| Dichiarazioni di pulizia | Free of RNase, DNase, DNA, ATP, and endotoxin |
| Formato | In rack |
| Stile punta | Automation |
| Imballaggio | 96 tips/rack, 10 racks/pack, 5 packs/case |
| Vedi altri risultati |