missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BBL™ Cooked Meat Medium with Glucose, Hemin and Vitamin K
General-purpose medium for cultivation of obligate anaerobes, specifically Clostridium spp.
Marca: BD 295982
Requires : Ancillary culture media, reagents, quality control organisms and laboratory equipment
Questo articolo non è restituibile. Consulta la politica di reso
Descrizione
Tubes with tight caps should be opened carefully to avoid injury due to breakage of glass.
Specifica
CE, BS12, CMPH2, MCM9 | |
2 to 25°C | |
United States | |
Heart Tissue granules 98g/L, Peptic digest of Animal Tissue 20g/L, Dextrose 2g/L, Sodium Chloride 5g/L | |
10 | |
9 mL |
Brown | |
Homogenous Pellets | |
Cooked Meat Medium | |
Cultivation of obligate anaerobes, Clostridium species | |
Tube |
For in vitro diagnostic use.