missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BBL™ Mycoflask™ Modified Trichosel™ Broth Prepare Media
Used for the isolation and cultivation of Trichomonas species
Marca: BD 298323
Dettagli aggiuntivi : Peso : 0.00900kg
Descrizione
- With 5% Horse Serum
Tubes with tight caps should be opened carefully to avoid injury due to breakage of glass.
Specifica
ISO/DIS, 11014 | |
2°C to 8°C | |
United States | |
Pancreatic digest of Casein 12g/L, Beef extract 2g/L, Yeast extract 5g/L, L-Cysteine HCI 1g/L, Maltose 2g/L, Agar 1g/L, Chloramphenicol 0.1g/L, Methylene blue 3mg/L, Horse Serum 5% | |
Tube with Tight Cap |
Light Beige | |
Liquid | |
Trichosel Broth, Modified, with 5% Horse Serum | |
Used for the isolation and cultivation of Trichomonas species | |
10 mL |
For in vitro diagnostic use.