Learn More
BD BBL™ Mycoflask™ Todd Hewitt Broth Prepared Media
Used for the cultivation of beta-hemolytic streptococci
Marca: BD 221714
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Tubes with tight caps should be opened carefully to avoid injury due to breakage of glass.
Specifica
CE | |
Liquid | |
Todd Hewitt Broth | |
Cultivation of beta-hemolytic streptococci | |
5 mL |
2 to 25°C | |
United States | |
Heart infusion from (solids) 3.1g/L, Peptone 20g/L, Dextrose 2g/L, Sodium Chloride 2g/L, Sodium Phosphate 0.4g/L, Sodium Carbonate 2.5g/L | |
Tube with Tight Cap |
For in vitro diagnostic use.