Learn More
BD BBL™ Mycoflask™ Urease Test Broth (Stuart's) Prepared Media
Used for the differentiation of organisms, especially the Enterobacteriaceae, on the basis of urease production
Marca: BD 221719
22.23 EUR valido fino al 2024-10-18
Usa il codice promo "21642" per avere il tuo prezzo promozionale.
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Tubes with tight caps should be opened carefully to avoid injury due to breakage of glass.
Specifica
CE, AOAC, BAM, COMPF, SMD | |
2°C to 8°C | |
United States | |
Urea 200g/L, Monopotassium Phosphate 91g/L, Disodium Phosphate 95g/L, Yeast extract 1g/L, Phenol red 0.1g/L | |
Tube with Tight Cap |
Yellow | |
Liquid | |
Urease Test Broth | |
For differentiation of organisms, especially Enterobacteriaceae, on basis of Urease production | |
3 mL |
For in vitro diagnostic use.