missing translation for 'onlineSavingsMsg'
Learn More

BD BBL™ Prepared Media Sabouraud Dextrose Agar Deeps

Used in qualitative procedures for cultivation of pathogenic and nonpathogenic fungi (particularly dermatophytes)

Marca:  BD 296182

Codice prodotto. 15174172

  • 371.00€ / pezzi 100
Spedizione stimata: 23-01-2025
per visualizzare lo stock disponibile
Esplora altre offerte speciali
Aggiungi al carrello
Questo articolo non è restituibile. Consulta la politica di reso

Descrizione

Descrizione

  • Prepared media with chloramphenicol
  • Pour tubes, Grouping: Q through S products
  • Appearance: Light to medium tan cream to yellow; trace hazy
  • Reaction at 25°C, pH: 5.6 ± 0.2
  • Meets United States Pharmacopeia (USP), European Pharmacopoeia (EP) and Japanese Pharmacopoeia (JP)1-3 performance specifications
  • Volume: 20mL

TRUSTED_SUSTAINABILITY
Specifica

Specifica

CE, BAM, CCAM, CMPH2, COMPF, EP, JP, MCM9, USP
2°C to 8°C
United States
Pancreatic digest of Casein 5g/L, Peptic digest of Animal Tissue 5g/L, Dextrose 40g/L, Agar 15g/L
Pour Tube with Cap
Light Beige
Liquid
Sabouraud Dextrose Agar
For qualitative procedures for cultivation of pathogenic and nonpathogenic fungi (particularly dermatophytes)
10 mL
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali

For in vitro diagnostic Use