Learn More
BD Cell Viability Solution
Click to view available options
Quantità:
100 Tests
500 Tests
Dimensione della confezione:
Pezzo
Descrizione
- Used as a viability probe for methods of dead cell exclusion, based on a light scatter and uptake of 7-AAD as detected in FL3
- Does not require dilution
Flow Cytometry
Specifica
Specifica
| Content And Storage | 4°C |
| Da utilizzare con (applicazione) | Flow Cytometry |
| Quantità | 500 Tests |