Learn More
BD Cytometric Bead Array (CBA) Human CD62E (E-Selectin); Flex Set, Bead D9
Bead-based immunoassay capable of measuring human soluble CD62E (E-Selectin) in serum, plasma and cell culture supernatant samples
Marca:  BD 560419
1193.05 EUR valido fino al 2025-12-31
Usa il codice promo "25339" per avere il tuo prezzo promozionale.
Codice prodotto. 15800189
  • 1195.00€ / Pezzo
Spedizione stimata: 24-01-2025
per visualizzare lo stock disponibile
Esplora altre offerte speciali
Aggiungi al carrello
Includes:
  • Human soluble CD62E (E-Selectin), Standard
  • Human soluble CD62E (E-Selectin), PE Detection Reagent
  • Human soluble CD62E (E-Selectin), Capture Bead D9
Questo articolo non è restituibile. Consulta la politica di reso
Descrizione
Descrizione
Contains one vial each of Capture Bead and PE Detection Reagent and two vials of Standard. The Capture Bead and PE Detection Reagent components of this flex set have been formulated to a 50x concentration to ensure product performance when multiplexed. The Standard component is lyophilized and should be transferred to a 15mL polypropylene tube for reconstitution. When reconstituted in 4.0mL Assay Diluent, the standard has a protein concentration of 10,000 pg/mL.
  • Human reactivity was determined by testing samples with CBA Human Soluble CD62E (E-Selectin) Flex Set
  • Must be used in conjunction with CBA Human Soluble Protein Master Buffer Kit, flow cytometer, and FCAP Array Software
TRUSTED_SUSTAINABILITY
Specifica
Specifica
CD62E (E-Selectin)
Human soluble CD62E (E-Selectin), standard, Human soluble CD62E (E-Selectin), PE detection reagent, Human soluble CD62E (E-Selectin), capture bead D9
4°C
Human CD62E (E-Selectin) flex set (bead D9)
Human
100 Tests
Flow Cytometry