Learn More
BD Cytometric Bead Array (CBA) Human Granzyme; Flex Set, Bead D9
Bead-based immunoassay capable of measuring soluble human Granzyme A in serum, plasma, and cell culture supernatant samples
Marca: BD 560299
Dettagli aggiuntivi : Peso : 0.04400kg
Includes:
- Human Granzyme A, Standard
- Human Granzyme A, PE Detection Reagent
- Human Granzyme A, Capture Bead D9
Descrizione
Contains one vial each of Capture Bead and PE Detection Reagent and two vials of Standard. The Capture Bead and PE Detection Reagent components of this flex set have been formulated to a 50x concentration to ensure product performance when multiplexed. The Standard component is lyophilized and should be transferred to a 15mL polypropylene tube for reconstitution. When reconstituted in 4.0mL Assay Diluent, the standard has a protein concentration of 10,000 pg/mL.
- Human reactivity was determined by testing samples with BA Human Granzyme A Flex Set
- Must be used in conjunction with CBA Human Soluble Protein Master Buffer Kit, flow cytometer, and FCAP Array Software
Specifica
Granzyme B | |
Human Granzyme A, standard, Human Granzyme A, PE detection reagent, Human Granzyme A, capture bead D9 | |
4°C | |
Human Granzyme A flex set (bead D9) |
Human | |
100 Tests | |
Flow Cytometry |