Learn More
BD Cytometric Bead Array (CBA) Human IL-1β; Flex Set, Bead B4
Bead-based immunoassay capable of measuring human interleukin-1Glc beta (IL-1Glc beta) in serum, plasma, and cell culture supernatant samples
Marca: BD 558279
Includes: Human IL-1β; Standard; Human IL-1β; PE Detection Reagent; Human IL-1β; Capture Bead B4
Descrizione
Reactivity
- Qc Testing: Human
Flow cytometry
Specifica
IL-1beta | |
Human IL-1beta, standard, Human IL-1beta, PE detection reagent, Human IL-1beta, capture bead B4 | |
Serum, Plasma, Cell Culture Supernatant | |
Flow Cytometry |
Human | |
100 Tests | |
4°C | |
Human IL-1beta flex set (bead B4) |