Learn More
BD Cytometric Bead Array (CBA) Mouse IL-21; Flex Set, Bead B6
Determines mouse reactivity by testing samples with BD CBA Mouse IL-21 Flex Set
Marca: BD 560160
824.97 EUR valido fino al 2025-12-31
Usa il codice promo "25339" per avere il tuo prezzo promozionale.
Includes: Mouse IL-21; Standard; Mouse IL-21; PE Detection Reagent; Mouse IL-21; Capture Bead B6
Descrizione
The BD™ CBA Mouse IL-21 Flex Set is a bead-based immunoassay capable of measuring mouse interleukin-21 (IL-21) in serum and cell culture supernatant samples
- Reactivity: QC Tested: Mouse
- Bead Position: B6
- Limit of detection is 4.8pg/mL
With BD CBA Mouse/Rat soluble protein master buffer kit (Cat. No. 558266, 100 tests, or 558267, 500 tests), flow cytometer, and FCAP Array™ software
Specifica
IL-21 | |
Mouse IL-21, standard, Mouse IL-21, PE detection reagent, Mouse IL-21, capture bead B6 | |
Flow Cytometry |
Mouse | |
100 Tests | |
Mouse IL-21 flex set (bead B6) |