Learn More
BD Human C5a ELISA Kit II
For the in vitro quantitative determination of Human C5a-desArg in human EDTA plasma, serum and other biological samples
Marca: BD 557965
Dettagli aggiuntivi : Peso : 0.10000kg
Includes:
Antibody Coated Wells, Detection Antibody, Standards, Enzyme Concentrate (250x), Standard/Sample Diluent, ELISA Diluent, Wash Concentrate (20x), TMB One-Step Substrate Reagent, Stop Solution, Plate Sealers
Descrizione
Induces the release of pharmacologically active mediators from a number of cell types
- Standard Range: 78-5000pg/mL
ELISA
Specifica
C5a | |
Stop solution, ELISA diluent, TMB one-step Substrate reagent, detection Antibody Biotin Anti-Human C5a, Human C5a standard, wash concentrate (20X), standard/Sample Diluent, Human C5a Antibody coated plate, Enzyme concentrate Streptavidin HRP | |
Human EDTA Plasma, Serum, Biological Sample | |
In-vitro quantitative determination of Human C5a-desArg, ELISA |
Human | |
1 Plate | |
2°C to 8°C | |
Human C5a ELISA kit II |
Research Use Only