missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Human MCP-1 ELISA Set
For natural or recombinant human MCP-1 in serum, plasma, and cell culture supernatants
Marca: BD 555179
Allerta:
Reagents which contain preservatives may be toxic if ingested, inhaled, or in contact with skin
Includes:
- Capture Antibody: Anti-human MCP-1 monoclonal antibody
- Detection Antibody: Biotinylated anti-human MCP-1 monoclonal antibody
- Enzyme Reagent: Streptavidin-horseradish peroxidase conjugate (SAv-HRP)
- Standards: Recombinant human MCP-1, lyophilized
Requires : 96-well BD Falcon™ ELISA plates (Cat. No. 353279) are recommended, Microplate reader capable of measuring absorbance at 450 nm, Precision pipettes, Graduated cylinder, one liter, Deionized or distilled water, Wash bottle or automated washer, Log-log graph paper or automated data reduction, Tubes to prepare standard dilutions, Laboratory timer, Plate sealers or parafilm
Questo articolo non è restituibile. Consulta la politica di reso
Descrizione
The OptEIA™ set for human monocyte chemoattractant protein-1(MCP-1) contains the components necessary to develop enzymelinked immunosorbent assays
- Standard Range: 7.8 - 500pg/mL
- Calibrated against purified Baculovirus-expressed recombinant human MCP-1
- Allow flexible assay design to fit individual laboratory needs
- To design an immunoassay with different sensitivity and dynamic range
ELISA
Specifica
7.8 to 500pg/mL | |
20 96-well Plates | |
anti-Human MCP-1 monoclonal (capture) Antibody, Biotinylated anti-Human MCP-1 monoclonal (detection) Antibody, Instruction/analysis certificate, Lyophilized recombinant Human MCP-1 (standard), Streptavidin-HRP Conjugate | |
2°C to 8°C |
Human MCP-1 ELISA set | |
To develop ELISA for natural or recombinant Human MCP-1 in Serum, Plasma and cell culture supernatants | |
20 Plates |
For research use only