Learn More
BD Microtainer™ MAP Microtube for Automated Process with K2EDTA and BD Microgard™ Closure
Designed for the collection, transport and automated processing of skin puncture samples from infants, children, geriatric, oncology and critical care patients
Marca: BD 363706
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
The BD Microtainer MAP tube is designed to improve turnaround time, safety and accuracy of capillary blood collection and testing in patients with limited blood volume such as infants, children, oncology patients and the elderly.
- Streamlines operations by allowing for automated processing of your micro-collection hematology samples
- Enhances patient safety by accommodating full-size patient identification labels at the time of collection
- Improves workflow with one-piece design, no parts to assemble, no contaminated parts to discard
- Latex-free, disposable and nonsterile
Specifica
Clear | |
1.0 mg K2 EDTA | |
250 to 500 μL | |
Lavender |
MAP MicroTube for Automated Process | |
Non-sterile | |
Yes |