missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Mouse IL-6 ELISA Set
For immunoassay protocols
Marca: BD 555240
Includes:
- Capture Antibody: anti-mouse IL-6 monoclonal antibody
- Detection Antibody: Biotinylated anti-mouse IL-6 monoclonal antibody
- Enzyme Reagent: Streptavidin-horseradish peroxidase conjugate (SAv-HRP)
- Standards: Recombinant mouse IL-6, lyophilized
- Instruction/Analysis Certificate
Descrizione
- Contains components necessary to develop enzyme-linked immunosorbent assays (ELISA)
- For natural or recombinant mouse IL-6 in serum, plasma, and cell culture supernatants
Enzyme Linked Immunosorbent Assay
Specifica
Mouse IL-6 ELISA Set | |
To develop ELISA for natural or recombinant Mouse IL-6 in serum, plasma and cell culture supernatants | |
2°C to 8°C |
Twenty 96-well Plates | |
anti-Mouse IL-6 monoclonal (capture) antibody, Biotinylated anti-Mouse IL-6 monoclonal (detection) antibody, Instruction/analysis certificate, Lyophilized recombinant Mouse IL-6 (standard), Streptavidin-HRP Conjugate |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Research Use Only
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto