missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD OptEIA™ Human IL-12 (p70) ELISA Set
For human interleukin-12 (p70) (IL-12 (p70))
Marca: BD 555183
Dettagli aggiuntivi : Peso : 0.04000kg
Descrizione
- Develops ELISA for natural or recombinant human IL-12 (p70) in serum, plasma, and cell culture supernatants
- Capture antibody: anti--human IL-12 (p70) monoclonal antibody
- Detection antibody: biotinylated anti-human IL-12 (p70) monoclonal antibody
- Enzyme reagent: Streptavidin-horseradish peroxidase conjugate
- Standards: lyophilized recombinant human IL-12 (p70)
- Standard range: 7.8 - 500pg/mL
ELISA
Specifica
7.8 to 500pg/mL | |
20 96-well Plates | |
anti-Human IL-12 (p70) monoclonal (capture) Antibody, Biotinylated anti-Human IL-12 (p70) monoclonal (detection) Antibody, Instruction/analysis certificate, Lyophilized recombinant Human IL-12 (p70), Streptavidin-HRP Conjugate | |
2°C to 8°C |
Human IL-12 (p70) ELISA set | |
To develop ELISA for natural or recombinant Human IL-12 (p70) in Serum, Plasma and cell culture supernatants | |
20 Plates |
For research use only.