Learn More
BD OptEIA™ Monkey IFN-γ ELISA Set
For natural or recombinant Interferon-γ (IFN-γ) in non-human primate (rhesus, cynomolgus, and baboon) serum, plasma and cell culture supernatants.
Brand:  BD 551492
Product Code. 15886668
  • 2105.00€ / Pezzo
Spedizione stimata: 28-11-2024
per visualizzare lo stock disponibile
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
Standard in this set is calibrated against purified Baculovirus-expressed recombinant human IFN-γ. One monkey IFN-γ unit (U) is defined as one picogram of recombinant human IFN-γ.
  • Kit contents: capture antibody (Anti-Monkey IFN-γ monoclonal antibody), detection antibody (Biotinylated Anti-Monkey IFN-γ monoclonal antibody), enzyme reagent (Streptavidin-horseradish peroxidase conjugate SAv-HRP), standards (Recombinant IFN-γ, lyophilized), instruction/analysis certificate
  • Size: 20 tests of 96-well format
ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Monkey IFN-g ELISA set
To develop ELISA for natural or recombinant IFN-g in non-Human Primate (rhesus,cynomolgus and baboon) Serum, Plasma and cell culture supernatants
20 Plates
20 96-well Plates
anti-Monkey IFN-g monoclonal (capture) antibody, Biotinylated anti-Monkey IFN-g monoclonal (detection) antibody, Instruction/analysis certificate, Lyophilized recombinant IFN-g (standard), Streptavidin-HRP
2°C to 8°C
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers