Learn More
BD Rat IL-4 ELISA Set
For rat interleukin-4 (IL-4)
Marca: BD 555198
Dettagli aggiuntivi : Peso : 0.16000kg
Descrizione
The OptEIA™ set contains components necessary to develop enzyme-linked immunosorbent assays (ELISA) for natural or recombinant rat IL-4 in serum, plasma, and cell culture supernatants.
- Capture antibody: anti-rat IL-4 monoclonal antibody
- Detection antibody: biotinylated anti-rat IL-4 monoclonal antibody
- Enzyme reagent: Streptavidin-horseradish peroxidase conjugate
- Standards: lyophilized recombinant rat IL-4
- Standard range: 1.6 - 100pg/mL
ELISA
Specifica
1.6 to 100pg/mL | |
20 96-well Plates | |
anti-Rat IL-4 monoclonal (capture) antibody, Biotinylated anti-Rat IL-4 monoclonal (detection) antibody, Instruction/analysis certificate, Lyophilized recombinant Rat IL-4 (standard), Streptavidin-HRP Conjugate | |
2°C to 8°C |
Rat IL-4 ELISA set | |
To develop ELISA for natural or recombinant Rat IL-4 in Serum, Plasma and cell culture supernatants | |
20 Plates |
For research use only.