Learn More
BD Recomninant Rat GM-CSF
Cytokine having potent stimulatory effects on growth and differentiation of bone marrow progenitor cells
Marca: BD 555111
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
- Produced by activated T cells, macrophages, vascular endothelial cells and fibroblast
- Acts on mature leukocytes to promote inflammatory responses in peripheral tissues
- Promotes inflammatory responses
- Storage buffer: frozen aqueous buffered solution containing BSA and glycerol
ELISA standard, bioassay
Specifica
-80°C | |
5 μ g | |
Rat |
For research use only.