Learn More
BD Vacutainer™ Pronto™ Quick Release Needle Holder
Designed to prevent over-tightening when needle is secured
Marca: BD 368872
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- Reusable plastic holder (100 estimated uses per holder), designed for use with all BD Vacutainer single and multiple sample needles, blood collection sets and Luer adapter
- Designed to prevent overtightening when needle is secured and to facilitate a smooth and trouble-free release
- Requires no assembly and can be used with any sharps container
Specifica
Prevent over-tightening when needle is secured | |
BD Vacutainer single and multiple sample needles, blood collection sets and luer adapter | |
Plastic | |
Quick Release Needle Holder |