Learn More
Sartorius Benchtop Arium pro UF Ultrapure Water System
Sfoglia Tutto Sartorius Prodotti
Click to view available options
:
Flow Rate: 1.7L/min.
Descrizione
- Modular units pre-assembled according to specific applications
- Lowest TOC level, along with real-time monitoring function
- Efficient removal of endotoxins, DNase and RNase
- High Flexibility
- Total ease of operation
- Meets and exceeds the ASTM Type 1 standard, ensuring reproducibility of results
- Applications: HPLC, GC-MS, AAS, ICP-MS, Ion chromatography, TOC analysis, PCR, Electrophoresis, Endotoxin analysis, Immunocytochemistry, Pure water for preparation of cell culture media, Pure water for production of monoclonal antibodies, Photometry
- 5000 MWCO microporous membrane retains macromolecules and the water can be used for cell culture applications
- Typical applications include cell cultivation, DNA sequence analysis, IVF and general biochemical appilcations
Specifica
Specifica
| Conducibilità | 100μS/cm |
| Contenuto batterico | <0.1 CFU/mL (with a 0.2 μm final filter or terminal ultrafiltration cartridge ) |
| Produzione quotidiana | 100 L |
| Concentrazione di endotossine | <0.001 EU/mL with a terminal ultrafiltration cartridge |
| Feedwater Connector | 3/8 in. |
| Controllo dell'acqua di alimentazione | No |
| Sorgente acqua di alimentazione | Distilled, DI, RO, EDI |
| Portata | 1.7 L/min. |
| Tipo di acqua prodotto | Type 1 |
| Erogatore a distanza | No |
| Vedi altri risultati |