missing translation for 'onlineSavingsMsg'
Learn More

Benzyl mercaptan, polymer supported, 1% cross-linked, 100-200 mesh, 2.0-4.0 mmol/g on poly(styrene-divinylbenzene), Thermo Scientific Chemicals

Catalog Number p-7059358
Click to view available options
Quantità:
2 g
10 g
50 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
100-53-8
C7H8S
124.20
MFCD00004867,MFCD00801588,MFCD01863867
UENWRTRMUIOCKN-UHFFFAOYSA-N
benzyl mercaptan, benzenemethanethiol, benzylthiol, alpha-toluenethiol, thiobenzyl alcohol, benzyl hydrosulfide, alpha-toluolthiol, benzylhydrosulfide, phenylmethyl mercaptan, benzylmercaptan
7509
phenylmethanethiol
SCC1=CC=CC=C1
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
100-53-8
C7H8S
124.20
MFCD00004867,MFCD00801588,MFCD01863867
UENWRTRMUIOCKN-UHFFFAOYSA-N
benzyl mercaptan, benzenemethanethiol, benzylthiol, alpha-toluenethiol, thiobenzyl alcohol, benzyl hydrosulfide, alpha-toluolthiol, benzylhydrosulfide, phenylmethyl mercaptan, benzylmercaptan
7509
phenylmethanethiol
SCC1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Materiale o nome chimico Benzyl mercaptan
Nota nome Polymer supported, 1% cross-linked, 2.0 to 4.0 mmol/g on poly(styrene-divinylbenzene)
Colore Pink to White
Dimensioni maglia 100 to 200 Mesh
Quantità 2 g
Forma fisica Beads

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'