Learn More
IBA Lifesciences BioLock
BioLock is a Biotin blocking solution for lysates and culture supernatants
213.00€ - 830.00€
Codice del prodotto | Marca | Capacità (sistema metrico) | Prezzo | Quantità e disponibilità | |||||
---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Capacità (sistema metrico) | Prezzo | Quantità e disponibilità | |||||
15643608
|
IBA Lifesciences
2-0205-050 |
50 mL |
213.00€
50mL |
In stock
Eseguire il login per visualizzare lo stock disponibile |
|||||
15653608
|
IBA Lifesciences
2-0205-250 |
250 mL |
830.00€
250mL |
Spedizione stimata: 06-09-2024
Eseguire il login per visualizzare lo stock disponibile |
|||||
Descrizione
BioLock is a ready-to-use biotin blocking solution which contains the tetrameric biotin binding protein avidin. BioLock prevents biotin and biotinylated proteins from binding to Strep-Tactin™ or Strep-Tactin™XT, whereas binding of the Twin-Strep-tag™ and Strep-tag™II is not influenced.Biotin and/or biotinylated proteins can possibly reduce the binding capacity of Strep-Tactin™ as well as Strep-Tactin™XT and are present in cell lysates and cell culture media, especially from mammalian or insect cells. Thus, if Twin-Strep-tag™ and Strep-tag™II fusion proteins from biotin-containing extracts or media are intended to be purified via Strep-Tactin™ or Strep-Tactin™XT affinity chromatography, biotin has to be masked by addition of BioLock prior to application onto the resin.
Specifica
Activity: >70 U/ mL | |
Blocking Solution |
For Research Use Only.