Learn More
ACROBiosystems Biotinylated Human NKG2D / CD314 Protein, His, Avitag
Marca:  ACROBiosystems NKD-H82E3-200ug
Dettagli aggiuntivi : Peso : 0.00010kg
Codice prodotto. 17286340
  • 2103.34€ / 200µg
Spedizione stimata: 11-09-2024
per visualizzare lo stock disponibile
Descrizione
Descrizione
NKG2D is a transmembrane protein belonging to the CD94/NKG2 family of C-type lectin-like receptors, also known as KLRK1, CD314, D12S2489E, KLR and killer cell lectin like receptor K1. NKG2D itself forms a homodimer whose ectodomains serve for ligand binding. NKG2D is a major recognition receptor for the detection and elimination of transformed and infected cells as its ligands are induced during cellular stress, either as a result of infection or genomic stress such as in cancer. In NK cells, NKG2D serves as an activating receptor, which itself is able to trigger cytotoxicity. The function of NKG2D on CD8+ T cells is to send co-stimulatory signals to activate them.
This protein carries a polyhistidine tag at the N-terminus, followed by an Avi tag (AvitagTM). The protein has a calculated MW of 20.2 kDa.
Specifica
Specifica
20.2 kDa
Biotinylated Human NKG2D, His,Avitag (NKD-H82E3) is expressed from human 293 cells (HEK293). It contains AA Ile 73 - Val 216 (Accession # P26718-1).
-20°C
NKG2D
Human
HEK293
MABSol™ Biotin Labeled Proteins
200 μg
Ile 73 - Val 216
NKG2D,CD314,KLRK1,NK cell receptor D
Precision Avi
His Tag & Avi Tag
Powder