Learn More
Bis(pyridine)iodonium tetrafluoroborate, 97%, Thermo Scientific Chemicals
70.80€ - 224.00€
Identificatori chimici
CAS | 15656-28-7 |
---|---|
Formula molecolare | C10H10BF4IN2 |
Molecular Weight (g/mol) | 371.91 |
Numero MDL | MFCD03703393 |
InChI Key | BMDSRCBKJZCUBH-UHFFFAOYSA-N |
Sinonimo | bis pyridine iodonium tetrafluoroborate, barluenga's reagent, bis pyridine iodonium i tetrafluoroborate, iodanium pyridine tetrafluoroborate, iodonium pyridine tetrafluoroborate, bispyridine iodonium tetrafluoroborate, bis pyridin iodonium tetrafluoroborate, iodium bis pyridine tetrafluoroborate, bis pyridine-iodonium tetrafluoroborate |
PubChem CID | 10883201 |
IUPAC Name | iodanium;pyridine;tetrafluoroborate |
SMILES | F[B-](F)(F)F.[I-]([N+]1=CC=CC=C1)[N+]1=CC=CC=C1 |
Codice del prodotto | Marca | Quantità | Imballaggio | Prezzo | Quantità e disponibilità | ||||
---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Quantità | Imballaggio | Prezzo | Quantità e disponibilità | ||||
10723993
|
Thermo Scientific Acros
369680010 |
1 g | Glass bottle |
70.80€
1g |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | ||||
10218753
|
Thermo Scientific Acros
369680050 |
5 g | Glass bottle |
224.00€
5g |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | ||||
Descrizione
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagentIdentificatori chimici
15656-28-7 | |
371.91 | |
BMDSRCBKJZCUBH-UHFFFAOYSA-N | |
10883201 | |
F[B-](F)(F)F.[I-]([N+]1=CC=CC=C1)[N+]1=CC=CC=C1 |
C10H10BF4IN2 | |
MFCD03703393 | |
bis pyridine iodonium tetrafluoroborate, barluenga's reagent, bis pyridine iodonium i tetrafluoroborate, iodanium pyridine tetrafluoroborate, iodonium pyridine tetrafluoroborate, bispyridine iodonium tetrafluoroborate, bis pyridin iodonium tetrafluoroborate, iodium bis pyridine tetrafluoroborate, bis pyridine-iodonium tetrafluoroborate | |
iodanium;pyridine;tetrafluoroborate |
Specifica
Bis(pyridine)iodonium tetrafluoroborate | |
137.0°C to 141.0°C | |
Authentic | |
C10H10BF4IN2 | |
15, 1303 | |
BMDSRCBKJZCUBH-UHFFFAOYSA-N | |
iodanium;pyridine;tetrafluoroborate | |
10883201 | |
97% |
15656-28-7 | |
Yellow | |
97% | |
MFCD03703393 | |
bis pyridine iodonium tetrafluoroborate, barluenga's reagent, bis pyridine iodonium i tetrafluoroborate, iodanium pyridine tetrafluoroborate, iodonium pyridine tetrafluoroborate, bispyridine iodonium tetrafluoroborate, bis pyridin iodonium tetrafluoroborate, iodium bis pyridine tetrafluoroborate, bis pyridine-iodonium tetrafluoroborate | |
F[B-](F)(F)F.[I-]([N+]1=CC=CC=C1)[N+]1=CC=CC=C1 | |
371.91 | |
371.91 | |
Powder |
Sicurezza e movimentazione
GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
- Bis(pyridine)iodonium tetrafluoroborate
Avvertenza
- Attenzione
Categoria di pericolo
- Lesione oculare grave/irritazione oculare Categoria 2
- Corrosione/irritazione della pelle Categoria 2
- Tossicità specifica per organi bersaglio (esposizione singola) Categoria 3
Indicazioni di pericolo
- H319-Provoca grave irritazione oculare.
- H335-Può irritare le vie respiratorie.
- H315-Provoca irritazione cutanea.
Consigli di prudenza
- P261-Evitare di respirare la polvere/i fumi/i gas/la nebbia/i vapori/gli aerosol.
- P280-Indossare guanti/indumenti protettivi/Proteggere gli occhi/il viso.
- P302+P352-IN CASO DI CONTATTO CON LA PELLE: lavare abbondantemente con acqua/.
- P305+P351+P338-IN CASO DI CONTATTO CON GLI OCCHI: sciacquare accuratamente per parecchi minuti. Togliere le eventuali lenti a contatto se è agevole farlo. Continuare a sciacquare.
RUO – Research Use Only