missing translation for 'onlineSavingsMsg'
Learn More

SHOWA™ Blue Chemical Protection Gloves, Model CS701

Catalog Number 17857351
Click to view available options
Size:
7
8
9
10
11
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

The CS701 food-safe gloves are designed with precision in mind. Anti-slip, rough surface grip gives the wearer ice-grip and optimum performance.

  • A double-dipped nitrile coating also provides chemical protection and durability to ensure safety from dangerous substances.
  • The seamless liner and natural latex-free composition are skin-friendly and comfortable for long-wear.
Chlorinated No
Colore Blue
Finger Thickness (English) 45.6 mil
Finger Thickness (Metric) 1.16 mm
Latex Free Yes
Materiale Nitrile, Polyester
Palm Thickness (English) 45.6 mil
Palm Thickness (Metric) 1.16 mm
Size 7
Spessore (sistema imperiale) 45.6 mil
Spessore (sistema metrico) 1.16 mm
EN388 Standard EN388:3132A
EN 421 No
EN ISO 374-1 Type A
EN ISO 374-1 CHEMICAL LETTER CODES JKLMNOPT
EN ISO 374 - 5 Bacteria, Fungi only
Stile polso guanto Gauntlet
Rivestimento interno Lined
Vedi altri risultati Mostra meno risultati
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'