Learn More
Emma Safety Footwear Bologna Brogue-Style Business Shoes
With ankle support in the shank.
Marca: Emma Safety Footwear MM10709046
Dettagli aggiuntivi : Peso : 0.50000kg
Descrizione
- Polished black full-grain leather uppers
- Impenetrable nitrile rubber sole
- Hydro-Tec™ sanitized silver and wellness leather inlay sole
- Woven zero penetration insole
- Anti-bacterial with leather top layer in-lay sole
- Antistatic properties: ESD 0,1 - 35 M Ohm / NEN-EN-IEC 61340-5-1
Specifica
Black | |
Ankle support | |
Anti-bacterial with leather top layer inlay sole | |
Brogue | |
Business Shoe |
Leather | |
46 | |
Impenetrable nitrile rubber | |
EN ISO 20345:2011; ESD 0,1-35 M Ohm/NEN-EN-IEC 61340-5-1 |