Learn More
Thermo Scientific™ Bolt™ Bis-Tris Plus XCell SureLock™ Mini-Cell Welcome Pack, 4-12%
The Bolt Welcome Pack provides all of the necessary Bolt gels and buffers needed to begin using the XCell SureLock Mini-Cell.
Marca: Thermo Scientific™ NW0412D
531.36 EUR valido fino al 2024-12-31
Usa il codice promo "21651" per avere il tuo prezzo promozionale.
Descrizione
The Bolt Welcome Pack provides all of the necessary Bolt gels and buffers needed to begin using the XCell SureLock Mini-Cell. The XCell SureLock Mini-Cell is a vertical mini-protein gel electrophoresis system compatible with all Invitrogen mini precast gels and Surecast handcast gels.
The Welcome Pack contains:
• XCell SureLock Mini-Cell (EI0001)
• Bolt Bis-Tris Plus gels (2 boxes, 20 gels)
• Bolt MES Running Buffer, 20X (B0002)
• Bolt LDS Sample Buffer, 4X (B0007)
• Bolt Sample Reducing Agent, 10X (B0009)
• PageRuler Plus Prestained Protein Ladder, 10 to 250 kDa (26619)
About the XCell SureLock Mini-Cell
The XCell SureLock Mini-Cell is a vertical mini-protein gel electrophoresis system compatible with all Invitrogen mini precast gels and Surecast handcast gels. The Mini-Cell is simple, sturdy, and easy to use. Each XCell SureLock Mini-Cell can accommodate up to two gels per run. Run times may vary depending on buffer conditions and power supplies used.
About Bolt Bis-Tris Plus Gels
Bolt Bis-Tris Plus gels are precast polyacrylamide gels designed for optimal separation of your small- to medium-sized proteins under denaturing conditions. Bolt Bis-Tris Plus gels are designed to deliver consistent gel performance and provide a neutral pH environment that minimizes protein modifications. Bolt gels are ideal for western blot transfer and analysis, and any other techniques where protein integrity is crucial. Also use Bolt gels to obtain optimal results for your day-to-day protein separation needs. Bolt Bis-Tris Plus gels come in four polyacrylamide concentrations: 8, 10, 12, and 4-12% gradient and multiple well formats. Bolt Bis-Tris gels feature wedge-shaped wells, which can easily load up to two times more sample volume allowing for detection of proteins in very dilute samples or visualization of low-abundance proteins.
For transferring your proteins to a membrane, we recommend using for Bolt Transfer Buffer (BT0006) for traditional wet transfer using the XCell II Blot Module (EI9051). Alternatively, rapid semi-dry transfer can be performed using the Invitrogen Power Blotter or rapid dry transfer using the iBlot 3 Gel Transfer Device (IB31001).
Specifica
XCell SureLock Mini-Cell | |
Mini | |
Bis-Tris | |
10-well | |
1 Welcome Pk. |
4 to 12% | |
1.0 mm | |
3.5 to 260 kDa | |
Bolt™ WedgeWell™ | |
Approved for shipment at room temperature and wet ice |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
For Research Use Only. Not for use in diagnostic procedures.