Learn More
Robu™ Büchner Filter Funnels with Porosity 3 VitraPOR™ Glass Filter Discs
Robu™ Büchner Filter Funnels with Glass Filter Discs are made of pure borosilicate 3.3 glass and are highly resistant to most chemicals, including solvents, acids, and alkalis. The VitraPOR™ filter disc is rated as Porosity 3 (Pore size: 16 to 40 μm, Standard: ISO 4793-80).
Marca:  Robu™ 21123
Dettagli aggiuntivi : Peso : 0.09000kg
Codice prodotto. 10022231
  • 66.40€ / Pezzo
Spedizione stimata: 04-09-2024
per visualizzare lo stock disponibile
Aggiungi al carrello
Descrizione
Descrizione
  • Product Type: Büchner Filter Funnel
  • Material: Borosilicate 3.3 Glass (Body), VitraPOR™ Glass (Filter)
  • Porosity: Grade 3
  • Pore Size: 16-40 μm
  • Standard: ISO 4793-80
  • For Use With (Application): Filtration
  • For Use With (Equipment): Guko Rubber Gaskets (e.g. 10430652)
Specifica
Specifica
65 mm (OD); 60 mm (ID)
10 mm
56 mm
Filtration
Guko Rubber Gaskets (e.g. 10430652)
16 to 40 μm
Imbuto per filtrazione Buchner
125 ml
Vetro borosilicato
ISO 4793-80
65 mm
Grade 3
4.2 oz.