Learn More
Rogo Sampaic™ Class B Glass Burettes
Excellent for dispensing measured amounts of chemical solutions in the laboratory. Rogo Sampaic™ Class B Glass Burettes are manufactured from durable clear borosilicate glass.
Marca: Rogo Sampaic™ DND008
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- With funnel
- Straight glass faucet
- Permanent red graduation marks
- Corrosion-free and resistant to thermal shock
Specifica
10 mL | |
0.1 mL | |
Buret |
Borosilicate Glass | |
±0.05 mL | |
Class B |