Learn More
3M™ Bovine Total Milk Protein ELISA Kit, E96MLK
Intended for the detection of bovine total milk proteins in clean-in-place water (CIP) final rinse water, environmental swab samples, food ingredients and processed food products.
Marca: 3M™ 7100151331
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- Designed to detect both processed and unprocessed target allergen proteins.
- Test can accommodate environmental swab, CIP rinse water and food samples.
- A universal extraction protocol streamlines testing for multiple analytes, saving you reagents and valuable time.
Specifica
Test | |
96 Wells | |
50 Minutes |