Learn More
Brady™ Self-laminating Label Cartridge for BMP™21 Series Printers
Self-laminating Label Cartridge for use with BMP21/BMP21-LAB and BMP21-PLUS label printers.
Marca: Brady™ 110929
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- Abrasion and smudge resistant
- Moisture resistant and self extinguishing
- For Communications Cables, Panel Identification, Voice/Data Identification, Wire and Cable Marking
- Compatible with BBP72 Label Printer, BBP81 Label Printer, Brady 200MVP Plus, Brady 2461, Brady 300MVP Plus, Brady 300X-Plus II, Brady 3481, Brady 360X-Plus II, Brady 600X-Plus II, Brady 6441, BradyPrinter PR Plus Series Label Printer, BradyPrinter PR Plus 600, Tagus T200
Compliance
RoHS Compliant, UL Recognized
Specifica
Labeling Ribbon | |
BMP21-LAB Label Printer, BMP21-PLUS Label Printer, IDPAL Handheld Label Maker, LABPAL Handheld Label Maker, Brady M210 Label Printer, Brady M210-LAB Label Printer, Brady M211 Label Printer | |
Thermal Transfer | |
1.25 in. | |
31.75 mm | |
Matte | |
-70°C to +70°C |
Black on White | |
Self-laminating Vinyl | |
14 ft. | |
4.27 m | |
4.27 m x 31.75 mm (14 ft. x 1.25 in.) | |
-94°F to +158°F | |
UL Recognized to UL 2238, RoHS compliant to 2011/65/EU (Pass 2015/863) |