missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ BRANDplates cellGrade™ 96-Well, cellGrade Premium-Treated, Flat-Bottom Microplate
Color:Clear; Sterility:Sterile; Lid:With Lid
Marca: BRAND™ 782082
201.58 EUR valido fino al 2024-11-30
Usa il codice promo "21681" per avere il tuo prezzo promozionale.
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- Poly-D-Lysine-equivalent surface with analogous results regarding cell growth and morphology
- Optimized cell adhesion reduces cell damage from frequent washing
- Allows for serum-free and serum-reduced cultivation
- Physical/chemical treatment has long room temperature shelf life compared to biological coatings
- Sterilized by β-radiation to ISO 11137 and AAMI guidelines for a SAL of 10-6
- Meet requirements of Ph. Eur. and USP 29
- DNA-, DNase-, RNase-free
- Endotoxins are <0.01 EU/mL
- Orange alphanumeric embossing for easy well identification
Order Info
Packs of 50 individually wrapped plates with lids
Specifica
ISO 11137, USP 29 | |
With Lid | |
1/Pk., 50/Cs. | |
Premium | |
96 | |
cellGrade Premium | |
Flat Bottom (F-well) | |
50/Cs. |
Standard | |
8 x 12 | |
Clear | |
Polystyrene | |
Sterile | |
350 μL | |
Cultivation of cell lines with the highest demands on their environment | |
1/Pk. |