Learn More
Branson Ultrasonics™ SLP Cup Horn for Sonifiers SLPe, SLPt, 150E and 150T
3 in. diameter cup horn for the SLP Sonifier Cell Disruptor is ideal for processing single or multiple vials and test tubes with identical parameters.
Marca: Branson Ultrasonics™ 109-116-1760
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Enables sonification of very small volumes without horn coming into contact with sample
- Cup horn is attached to the inverted converter
- Inlet and outlet enables liquid to circulate within the chip horn, to reduce heat buildup and overflow
- Horn can also be used without liquid cooling circulation
- Process samples in complete isolation
- Ideal for infectious solutions
- Precludes possibility of any titanium residue
- Ideal for single or batch processing
- Ideal for very small volume processing
Cell Disruption, Liposome preparation, Protein shearing and extraction, Releasing cellular components, including DNA and RNA, Tissue culture Infectious solutions
Specifica
3 in. | |
Titanium | |
Sonifier SLP, 150E, 150T |
5 in. | |
Indirect method of disrupting |