Learn More
Hirschmann™ Bright Lined Counting Chambers
Codice prodotto. 10517831
Click to view available options
Product Type:
Counting Chamber (Burker)
Counting Chamber (Fuch-Rosenthal)
Counting Chamber (Neubauer)
Counting Chamber (Thoma)
Dimensione della confezione:
Pezzo
Codice prodotto. 10517831
Marca: Hirschmann™ 8100205
Includes 2 haemocytometer glass covers.
  • Double net ruling
  • Bright lined
  • With LOT number on packaging
  • Labelled with: usable until, CE pursuant to IVD 98/79 EC
Specifica
Product Type Counting Chamber (Thoma)
Profondità (sistema metrico) 0.1 mm
Certificazione/Conformità CE pursuant to IVD 98/79 EC
Include 2 haemocytometer glass covers