missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTX™ BTXPress™ Cytoporation Low Conductivity Medium T4
BTXPress™ Cytoporation Low Conductivity Medium T4 is an advanced electroporation buffer designed for use with the BTX™ AgilePulse™ MAX Large Volume Electroporation System for ex vivo or in vitro delivery of DNA, RNA, oligonucleotides, and siRNA.
Marca: BTX™ 470003
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- Sterile-filtered from the highest quality non-animal, medical-grade reagents
- Buffer can be directly diluted in complete growth media for post-electroporation cell culture
- Optimized for eukaryotic electrofusion applications
- Maximum transfection efficiency, high cell viability
- Low conductivity for minimal heating
- Physiological pH and balanced osmolarity
- Contains no animal products
- Certified RNase and DNase free
Specifica
500 mL Bottle | |
BTX AgilePulse MAX Large Volume Electroporation Systems | |
Electroporation Consumable | |
18 months from date of manufacture | |
Sterile | |
Square Wave |
Ex vivo or in vitro delivery of DNA, RNA, oligonucleotides, and siRNA | |
BTX Electroporation | |
BTXpress Cytoporation Low Conductivity Medium T4 | |
500 mL | |
2°C to 8°C |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto