Learn More
BUCHI Glass Condensers
Sleek cold-trap glass condenser
Marca:  BUCHI 040645
Dettagli aggiuntivi : Peso : 2.69000kg
Codice prodotto. 12045502
  • 2070.00€ / Pezzo
Spedizione stimata: 04-12-2024
per visualizzare lo stock disponibile
Aggiungi al carrello
Descrizione
Descrizione
Type C condenser is equipped with a cold-trap insert for cooling with dry ice.
Includes:
  • Condenser outer glass
  • Condenser inner
  • Screw caps
  • Hose connections
  • Rubber drip ring
Specifica
Specifica
Condenser outer glass, condenser inner, screw caps and hose connections, rubber drip ring
Type C condenser is equipped with a cold-trap insert for cooling with dry ice
Cold Trap Condenser