missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ C16orf9 Recombinant Protein
Marca: Abnova™ H00083986-P01.25ug
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Specifica
AAH13047 | |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
86.46 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
25 μg | |
MLDHKDLEAEIHPLKNEERKSQENLGNPSKNEDNVKSAPPQSRLSRCRAAAFFLSLFLCLFVVFVVSFVIPCPDRPASQRMWRIDYSAAVIYDFLAVDDINGDRIQDVLFLYKNTNSSNNFSRSCVDEGFSSPCTFAAAVSGANGSTLWERPVAQDVALVECAVPQPRGSEAPSACILVGRPSSFIAVNLFTGETLWNHSSSFSGNASILSPLLQVPDVDGDGAPDLLVLTQEREEVSGHLYSGSTGHQIGLRGSLGVDGESGFLLHVTRTGAHYILFPCASSLCGCSVKGLYEKVTGSGGPFKSDPHWESMLNATTRRMLSHSSGAVRYLMHVPGNAGADVLLVGSEAFVLLDGQELTPRWTPKAAHVLRKPIFGRYKPDTLAVAVENGTGTDRQILFLDLGTGAVLCSLALPSLPGGPLSASLPTADHRSAFFFWGLHELGSTSETETGEARHSLYMFHPTLPRVLLELANVSTHIVAFDAVLFEPSRHAAYILLTGPADSEAPGLVSVIKHKVRDLVPSSRVVRLGEGGPDSDQAIRDRFSRLRYQSEA | |
C16orf9/DKFZp761D0211/FLJ32603/gs19 | |
ITFG3 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Protein Array, ELISA, Western Blot | |
83986 | |
C16orf9 (Human) Recombinant Protein (P01) | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Wheat Germ (in vitro) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ITFG3 | |
Human | |
Recombinant | |
Solution |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto