missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ C22orf1 (Human) Recombinant Protein
Human C22orf1 full-length ORF ( AAH28035, 1 a.a. - 326 a.a.) recombinant protein with GST-tag at N-terminal.
Marca: Abnova™ H00000758-P01.25ug
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- Sequence: MWRSRWDASVLKAEALALLPCGLGMAFSQSHVMAARRHQHSRLIIEVDEYSSNPTQAFTFYNINQGRFQPPHVQMVDPVPHDAPKPPGYTRFVCVSDTHSRTDPIQMPYGDVLIHAGDFTELGLPSEVKKFNEWLGSLPYEYKIVIAGNHELTFDQEFMADLIKQDFYYFPSVSKLKPENYENVQSLLTNCIYLQDSEVTVRGFRIYGSPWQPWFYGWGFNLPRGQALLEKWNLIPEGVDILITHGPPLGFLDWVPKKMQRVGCVELLNTVQRRVQPRLHVFGHIHEGYGVMADGTTTYVNASVCTVNYQPVNPPIVIDLPTPRNS
Specifica
AAH28035 | |
metallophosphoesterase domain containing 1 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MPPED1 | |
GST |
758 | |
Wheat germ expression system | |
25 μg | |
239AB, C22orf1, FAM1A, MGC88045 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |