Learn More
Novus Biologicals™ C32 Nuclear Cell Lysate
Codice prodotto. 18516181 Sfoglia Tutto Bio Techne Prodotti
Click to view available options
Quantità:
0.05 mg
Dimensione della confezione:
50µg
Codice prodotto. 18516181
Marca: Novus Biologicals™ L002V4
Nuclear Extract Lysate
TRUSTED_SUSTAINABILITY
Specifica
Metodo di preparazione Use it directly for immuno-precipitation, or heat lysate with SDS gel loading buffer to 95C for 5 minutes followed by rapid cooling for western blot application. If dissociating conditions are required, add reducing agent prior to heating.
Quantità 0.05 mg
Content And Storage Store at -80C. Avoid freeze-thaw cycles.
Bersaglio C32