missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CA7 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA5103703
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Antibody detects endogenous levels of total CA7.
Carbonic anhydrases are a large family of zinc metalloenzymes that catalyze the reversible hydration of carbon dioxide. They participate in a variety of biological processes, including respiration, calcification, acid-base balance, bone resorption, and the formation of aqueous humor, cerebrospinal fluid, saliva, and gastric acid. They show extensive diversity in tissue distribution and in their subcellular localization. This gene encodes a glycosylphosphatidyl-inositol-anchored membrane isozyme expressed on the luminal surfaces of pulmonary capillaries and proximal renal tubules. Its exact function is not known; however, it may have a role in inherited renal abnormalities of bicarbonate transport.Specifica
CA7 | |
Polyclonal | |
Unconjugated | |
Ca7 | |
AV343731; CA7; Car7; Carbonate dehydratase VII; carbonic anhydrase 7; Carbonic anhydrase VII; carbonic dehydratase VII; CAVII; CA-VII | |
Rabbit | |
Affinity chromatography | |
RUO | |
12354, 291819, 766 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P43166, Q9ERQ8 | |
Ca7, Car7 | |
A synthesized peptide derived from human CA7(Accession P43166), corresponding to amino acid residues I212-F262. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto