Learn More
DiscoverX cAMP Hunter™ CHO-K1 CXCR3 Gi Cell Line
The cAMP Hunter™ CHO-K1 CXCR3 Gi Cell Line measures CXCR3 (GPCR) activity via decreased levels of intracellular cAMP. This cell line ships as two vials of cryopreserved cells.
Marca: DiscoverX 95-0054C2
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
The cAMP Hunter CXCR3 Gi cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. CXCL11/I-TAC) based activation of CXCR3 GPCR activity via detection of decreasing levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wildtype CXCR3 , a naturally Gi/Go coupled GPCR. Upon activation of CXCR3 , the decrease in levels of intracellular cAMP leads to a decreased signal from EFC when the cAMP detection kit is applied. This stable modified cell line is provided as two vials of cryopreserved cells.Specifica
Intracellular cAMP level | |
Cell Lines | |
2 Vials | |
AY242128 |
CHO-K1 | |
Recombinant Human I-TAC (CXCL11) | |
CXCR3 |