Learn More
DiscoverX cAMP Hunter™ CHO-K1 GABBR1-GABBR2 Gi Cell Line
The cAMP Hunter™ CHO-K1 GABBR1-GABBR2 Gi Cell Line measures GABBR1 (GPCR) activity via decreased levels of intracellular cAMP. This cell line ships as two vials of cryopreserved cells.
Marca: DiscoverX 95-0165C2
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
The cAMP Hunter GABBR1-GABBR2 Gi cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. GABA) based activation of GABBR1 GPCR activity via detection of decreasing levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wildtype GABBR1, a naturally Gi/Go coupled GPCR, alongside its partner subunit GABBR2. Upon activation of GABBR1 , the decrease in levels of intracellular cAMP leads to a decreased signal from EFC when the cAMP detection kit is applied. This stable modified cell line is provided as two vials of cryopreserved cells.Specifica
Intracellular cAMP level | |
Cell Lines | |
2 Vials | |
NM_021903.2 |
CHO-K1 | |
GABA | |
GABBR1-GABBR2 |