Learn More
Erlab™ Captair Smart Filtering Fume Hood Mobicap
Erlab™ Captair Smart Filtering Fume Hood Mobicap offers a mobile, rolling bench for Captair Smart fume hoods.
Marca: Erlab™ I0560000201
546.10 EUR valido fino al 2024-09-17
Usa il codice promo "21690" per avere il tuo prezzo promozionale.
Dettagli aggiuntivi : Peso : 30.00000kg
Descrizione
- For Captair Smart filtering fume hoods
- Equipped with 4 wheels and 2 self-locks
- Allows the fume hood to be safely moved
Specifica
Smart 321 Fume Hood | |
Without Work Surface | |
Metal |
890 mm | |
Mobile Rolling Cart Work Bench |