Learn More
Erlab™ Captair™ Store™ Filter Alarm
Marca: Erlab™ I0299000001
Dettagli aggiuntivi : Peso : 0.80000kg
Specifica
For solvents (M and XLS fume hoods and Captair™ Store™) | |
Filter Saturation Electronic Alarm Molecode S | |
For Auto Detection of Filter Saturation by Solvents |