Learn More
Thermo Scientific™ CaptureSelect™ Biotin Anti-C1-Inhibitor Conjugate
Thermo Scientific™ CaptureSelect™ Biotin Anti-C1-Inhibitor Conjugate consists of a 13 kDa recombinant single domain antibody fragment (VHH affinity ligand) that specifically binds with high affinity and selectivity to human C1 esterase inhibitor (C1-Inh).
514.00€
Specifica
Content And Storage | Store in freezer (-5°C to -30°C). |
---|---|
Label Type | Biotin and Analogs |
Linea di prodotti | CaptureSelect™ |
Target Species | Human |
Tipo di legante | Antibody |
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
18426738
|
Thermo Scientific™
7103402100 |
100 μg |
514.00€
100µg |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
Descrizione
CaptureSelect Biotin Anti-C1-Inhibitor Conjugate consists of a 13 kDa recombinant single domain antibody fragment (VHH affinity ligand) that specifically binds with high affinity and selectivity to human C1 esterase inhibitor (C1-Inh). The affinity ligand is chemically conjugated to biotin via an appropriate spacer that retains the binding reactivity of the ligand when used in combination with streptavidin-based conjugates or streptavidin pre-coated surfaces. This conjugate allows you to detect, quantitate, and characterize native and recombinant human C1 esterase inhibitor.
Applications for CaptureSelect Biotin Anti-C1-Inhibitor Conjugate include ELISA, immunoprecipitation with streptavidin-coupled DynaBeads, Gyrolab™-based immunoassays, and label-free detection using platforms such as those based on surface plasmon resonance (Biacore™ and IBIS-MX96 systems) and bio-layer interferometry (ForteBio™ Octet™ systems).
Animal origin statement
CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12–15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).
Specifica
Store in freezer (-5°C to -30°C). | |
CaptureSelect™ | |
Antibody |
Biotin and Analogs | |
Human |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.